Assay Details
| Gene Symbol: LOC101712006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013065916.2 | XP_012921370.1 | 2 - 3 | 367 | 78 | |
| XM_021245077.1 | XP_021100736.1 | 2 - 3 | 368 | 78 | ||
More Information
Set Membership: |
|

LOC101712006
Genomic Map