Assay Details
| Gene Symbol: Grsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068143.2 | XP_012923597.1 | 6 - 7 | 1192 | 73 | |
| XM_013068144.2 | XP_012923598.1 | 6 - 7 | 1193 | 73 | ||
More Information
Set Membership: |
|

Grsf1
Genomic Map