Assay Details
| Gene Symbol: Ribc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251234.1 | XP_021106893.1 | 3 - 4 | 482 | 63 | |
| XM_021251235.1 | XP_021106894.1 | 3 - 4 | 486 | 63 | ||
| XM_021251236.1 | XP_021106895.1 | 3 - 4 | 481 | 63 | ||
More Information
Set Membership: |
|

Ribc1
Genomic Map