Assay Details
| Gene Symbol: LOC101717595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838454.2 | XP_004838511.2 | 11 - 12 | 1577 | 70 | |
| XM_021252763.1 | XP_021108422.1 | 11 - 12 | 1577 | 70 | ||
More Information
Set Membership: |
|

LOC101717595
Genomic Map