Assay Details
| Gene Symbol: LOC101713453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848105.3 | XP_004848162.1 | 3 - 4 | 745 | 103 | |
| XM_021261099.1 | XP_021116758.1 | 3 - 4 | 261 | 103 | ||
More Information
Set Membership: |
|

LOC101713453
Genomic Map