Assay Details
| Gene Symbol: Ppp1r3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864173.2 | XP_004864230.1 | 1 - 2 | 3274 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r3e| Gene Symbol: Ppp1r3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864173.2 | XP_004864230.1 | 1 - 2 | 3274 | 99 | |
Set Membership: |
|