Assay Details
| Gene Symbol: Klk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866784.2 | XP_004866841.1 | 3 - 4 | 470 | 58 | |
| XM_004866785.2 | XP_004866842.1 | 3 - 4 | 467 | 58 | ||
More Information
Set Membership: |
|

Klk15
Genomic Map