Assay Details
| Gene Symbol: Shox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834511.2 | XP_004834568.1 | 1 - 2 | 895 | 75 | |
| XM_004834512.2 | XP_004834569.1 | 1 - 2 | 895 | 75 | ||
More Information
Set Membership: |
|

Shox2
Genomic Map