Assay Details
| Gene Symbol: Csf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836630.2 | XP_004836687.1 | 1 - 2 | 156 | 70 | |
| XM_021243228.1 | XP_021098887.1 | 1 - 2 | 156 | 70 | ||
More Information
Set Membership: |
|

Csf2
Genomic Map