Assay Details
| Gene Symbol: Tcp11l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844812.3 | XP_004844869.1 | 5 - 6 | 1000 | 62 | |
| XM_004844813.3 | XP_004844870.1 | 5 - 6 | 997 | 62 | ||
| XM_004844814.3 | XP_004844871.1 | 5 - 6 | 997 | 62 | ||
More Information
Set Membership: |
|

Tcp11l2
Genomic Map