Assay Details
| Gene Symbol: Tnfrsf10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848338.3 | XP_004848395.2 | 8 - 9 | 1209 | 62 | |
| XM_004848339.3 | XP_004848396.1 | 8 - 9 | 1210 | 62 | ||
| XM_013073426.2 | XP_012928880.1 | 9 - 10 | 1123 | 62 | ||
| XM_013073427.2 | XP_012928881.1 | 7 - 8 | 1098 | 62 | ||
| XM_021261277.1 | XP_021116936.1 | 7 - 8 | 1030 | 62 | ||
More Information
Set Membership: |
|

Tnfrsf10b
Genomic Map