Assay Details
| Gene Symbol: Zbtb26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262117.1 | XP_021117776.1 | 1 - 2 | 442 | 84 | |
| XM_021262118.1 | XP_021117777.1 | 1 - 2 | 441 | 84 | ||
| XM_021262119.1 | XP_021117778.1 | 1 - 2 | 442 | 84 | ||
| XM_021262120.1 | XP_021117779.1 | 1 - 2 | 441 | 84 | ||
More Information
Set Membership: |
|

Zbtb26
Genomic Map