Assay Details
| Gene Symbol: Creb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839496.2 | XP_004839553.1 | 7 - 8 | 856 | 65 | |
| XM_013070258.2 | XP_012925712.2 | 3 - 4 | 297 | 65 | ||
| XM_021254087.1 | XP_021109746.1 | 7 - 8 | 825 | 65 | ||
More Information
Set Membership: |
|

Creb5
Genomic Map