Assay Details
| Gene Symbol: Fkbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868143.2 | XP_004868200.1 | 1 - 2 | 316 | 81 | |
| XM_004868144.2 | XP_004868201.1 | 1 - 2 | 319 | 81 | ||
More Information
Set Membership: |
|

Fkbp7
Genomic Map