Assay Details
| Gene Symbol: Shcbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862835.2 | XP_004862892.2 | 2 - 3 | 556 | 69 | |
| XM_021242908.1 | XP_021098567.1 | 2 - 3 | 282 | 69 | ||
More Information
Set Membership: |
|

Shcbp1l
Genomic Map