Assay Details
| Gene Symbol: Tfpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872312.3 | XP_004872369.1 | 2 - 3 | 395 | 76 | |
| XM_021250261.1 | XP_021105920.1 | 2 - 3 | 141 | 76 | ||
| XM_021250262.1 | XP_021105921.1 | 2 - 3 | 141 | 76 | ||
| XM_021250263.1 | XP_021105922.1 | 2 - 3 | 141 | 76 | ||
More Information
Set Membership: |
|

Tfpi
Genomic Map