Assay Details
| Gene Symbol: Neu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247101.1 | XP_021102760.1 | 1 - 2 | 752 | 109 | |
| XM_021247102.1 | XP_021102761.1 | 2 - 3 | 723 | 109 | ||
More Information
Set Membership: |
|

Neu2
Genomic Map