Assay Details
| Gene Symbol: Med20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021259826.1 | XP_021115485.1 | 3 - 4 | 553 | 83 | |
| XM_021259828.1 | XP_021115487.1 | 3 - 4 | 2404 | 83 | ||
| XM_021259829.1 | XP_021115488.1 | 3 - 4 | 520 | 83 | ||
More Information
Set Membership: |
|

Med20
Genomic Map