Assay Details
| Gene Symbol: Eci2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847756.3 | XP_004847813.1 | 7 - 8 | 861 | 70 | |
| XM_004847757.3 | XP_004847814.2 | 7 - 8 | 829 | 70 | ||
More Information
Set Membership: |
|

Eci2
Genomic Map