Assay Details
| Gene Symbol: LOC101711916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851362.1 | XP_004851419.1 | 8 - 9 | 1259 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101711916| Gene Symbol: LOC101711916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851362.1 | XP_004851419.1 | 8 - 9 | 1259 | 65 | |
Set Membership: |
|