Assay Details
| Gene Symbol: Prdx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852507.3 | XP_004852564.2 | 2 - 3 | 387 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prdx5| Gene Symbol: Prdx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852507.3 | XP_004852564.2 | 2 - 3 | 387 | 60 | |
Set Membership: |
|