Assay Details
| Gene Symbol: Gna15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865822.2 | XP_004865879.2 | 5 - 6 | 1115 | 62 | |
| XM_021245073.1 | XP_021100732.1 | 5 - 6 | 1115 | 62 | ||
More Information
Set Membership: |
|

Gna15
Genomic Map