Assay Details
| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868906.3 | XP_004868963.1 | 3 - 4 | 387 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gypa| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868906.3 | XP_004868963.1 | 3 - 4 | 387 | 76 | |
Set Membership: |
|