Assay Details
| Gene Symbol: Abcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874948.3 | XP_004875005.1 | 5 - 6 | 2077 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcd1| Gene Symbol: Abcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874948.3 | XP_004875005.1 | 5 - 6 | 2077 | 59 | |
Set Membership: |
|