Assay Details
| Gene Symbol: Noxo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873676.3 | XP_004873733.1 | 3 - 4 | 400 | 114 | |
| XM_021251438.1 | XP_021107097.1 | 3 - 4 | 400 | 114 | ||
More Information
Set Membership: |
|

Noxo1
Genomic Map