Assay Details
| Gene Symbol: LOC101696295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021236468.1 | XP_021092127.1 | 2 - 3 | 2040 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101696295| Gene Symbol: LOC101696295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021236468.1 | XP_021092127.1 | 2 - 3 | 2040 | 75 | |
Set Membership: |
|