Assay Details
| Gene Symbol: Tubb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075272.2 | XP_012930726.1 | 1 - 2 | 1302 | 129 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013075272.2 transcripts.
Set Membership: |
|

Tubb6
Genomic Map