Assay Details
| Gene Symbol: LOC101708152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841945.2 | XP_004842002.1 | 1 - 2 | 462 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101708152| Gene Symbol: LOC101708152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841945.2 | XP_004842002.1 | 1 - 2 | 462 | 110 | |
Set Membership: |
|