Assay Details
| Gene Symbol: Slirp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837118.3 | XP_004837175.1 | 1 | 118 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004837118.3 transcripts.
Set Membership: |
|

Slirp
Genomic Map