Assay Details
| Gene Symbol: Vamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869392.3 | XP_004869449.1 | 5 | 2926 | 118 | |
| XM_021248132.1 | XP_021103791.1 | 5 | 2703 | 118 | ||
More Information
Set Membership: |
|

Vamp1
Genomic Map