Assay Details
| Gene Symbol: TLR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003266.3 | NP_003257.1 | 1 - 2 | 386 | 68 | |
| GenBank mRNA | AK226155.1 | - | 1 - 2 | 141 | 68 | |
| AK290053.1 | - | 1 - 2 | 369 | 68 | ||
| AL710219.1 | - | 1 - 2 | 236 | 68 | ||
| BC143841.1 | - | 1 - 2 | 108 | 68 | ||
| U88880.1 | - | 1 - 2 | 132 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000601.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK290053.1,NM_003266.3,U88880.1 transcripts.
Set Membership: |
|

TLR4
Genomic Map