Assay Details
| Gene Symbol: COX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303.3 | NP_001294.2 | 1 - 2 | 249 | 82 | |
| GenBank mRNA | AK312718.1 | - | 1 - 2 | 144 | 82 | |
| BC000060.2 | - | 1 - 2 | 122 | 3508180 | 82 | |
| BC006394.2 | - | 1 - 2 | 122 | 4136163 | 82 | |
| U09466.1 | - | 1 - 2 | 85 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000162.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312718.1,BC000060.2,BC006394.2,NM_001303.3,U09466.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial fission
mitochondrial electron transport, cytochrome c to oxygen
heme biosynthetic process
heme a biosynthetic process
respiratory chain complex IV assembly
aerobic respiration
cellular respiration
heme O biosynthetic process
hydrogen ion transmembrane transport
cytochrome-c oxidase activity
farnesyltranstransferase activity
protoheme IX farnesyltransferase activity
mitochondrial fission
mitochondrial electron transport, cytochrome c to oxygen
heme biosynthetic process
heme a biosynthetic process
respiratory chain complex IV assembly
aerobic respiration
cellular respiration
heme O biosynthetic process
hydrogen ion transmembrane transport
cytochrome-c oxidase activity
farnesyltranstransferase activity
protoheme IX farnesyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17002280.hg.1 | ENST00000261643.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002280.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002280.hg.1 | TC17000162.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002281.hg.1 | ENST00000261643.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002281.hg.1 | ENST00000536205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002281.hg.1 | ENST00000537334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002281.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002281.hg.1 | TC17000162.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002279.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002279.hg.1 | TC17000162.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002282.hg.1 | ENST00000261643.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002282.hg.1 | ENST00000536205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002282.hg.1 | ENST00000537334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002282.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002282.hg.1 | TC17000162.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700148897.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148897.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148897.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148897.hg.1 | TC1700006939.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148897.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148897.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148897.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148895.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148895.hg.1 | TC1700006939.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148895.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148899.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148899.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148899.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148899.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148899.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148899.hg.1 | TC1700006939.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148899.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148899.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148899.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148899.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148899.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148901.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148901.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148901.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148901.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148901.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148901.hg.1 | TC1700006939.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148901.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148901.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148901.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148901.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148901.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000261643.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000580561.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | ENST00000581931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006939.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148896.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148896.hg.1 | NM_001303.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700148896.hg.1 | TC1700006939.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700148896.hg.1 | ENST00000429152.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700148896.hg.1 | NM_001303.3 | Refseq |

COX10
Genomic Map