Assay Details
| Gene Symbol: PSMA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002792.3 | NP_002783.1 | 1 - 2 | 256 | 63 | |
| GenBank mRNA | AF022815.1 | - | 1 - 2 | 125 | 63 | |
| AF054185.1 | - | 1 - 2 | 186 | 63 | ||
| AK312025.1 | - | 1 - 2 | 151 | 63 | ||
| BC001895.1 | - | 1 - 2 | 189 | 63 | ||
| BC004427.1 | - | 1 - 2 | 174 | 2819923 | 63 | |
| BC014999.1 | - | 1 - 2 | 184 | 63 | ||
| BG575009.1 | - | 1 - 2 | 243 | 63 | ||
| BI258951.1 | - | 1 - 2 | 256 | 63 | ||
| BI906714.1 | - | 1 - 2 | 142 | 63 | ||
| BT007165.1 | - | 1 - 2 | 101 | 63 | ||
| GQ472224.1 | - | 1 - 2 | 174 | 63 | ||
| HY039591.1 | - | 1 - 2 | 256 | 63 | ||
More Information
Set Membership: |
|

PSMA7
Genomic Map