| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22004018.hg.1 |
ENST00000215838.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22004018.hg.1 |
ENST00000405742.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22004018.hg.1 |
ENST00000407817.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22004018.hg.1 |
NM_000355.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22004018.hg.1 |
NM_001184726.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22004018.hg.1 |
TC22000215.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
ENST00000215838.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
ENST00000405742.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
ENST00000407817.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
ENST00000450638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
NM_000355.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
NM_001184726.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
TC2200009257.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
ENST00000215838.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
ENST00000405742.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
ENST00000407817.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
ENST00000450638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
NM_000355.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200168526.hg.1 |
NM_001184726.1 |
Refseq |