Assay Details
| Gene Symbol: APOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000483.4 | NP_000474.2 | 1 - 2 | 99 | 86 | |
| GenBank mRNA | AV694494.1 | - | 1 - 2 | 95 | 86 | |
| BU596546.1 | - | 1 - 2 | 135 | 86 | ||
| Gene Symbol: APOC4-APOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037932.1 | 3 - 4 | 1199 | 86 | ||
| GenBank mRNA | AK126257.1 | - | 4 - 5 | 1133 | 86 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19002642.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_000483.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
retinoid metabolic process
positive regulation of phospholipase activity
positive regulation of triglyceride catabolic process
positive regulation of very-low-density lipoprotein particle remodeling
negative regulation of very-low-density lipoprotein particle clearance
lipid catabolic process
negative regulation of cholesterol transport
cholesterol efflux
phospholipid efflux
triglyceride-rich lipoprotein particle remodeling
chylomicron remodeling
very-low-density lipoprotein particle remodeling
chylomicron remnant clearance
high-density lipoprotein particle clearance
lipoprotein metabolic process
cholesterol homeostasis
negative regulation of catalytic activity
reverse cholesterol transport
positive regulation of fatty acid biosynthetic process
negative regulation of lipid metabolic process
negative regulation of receptor-mediated endocytosis
positive regulation of lipoprotein lipase activity
positive regulation of phospholipid catabolic process
triglyceride homeostasis
lipid binding
phospholipase activator activity
protein homodimerization activity
phospholipase binding
lipase inhibitor activity
lipoprotein lipase activator activity
retinoid metabolic process
positive regulation of phospholipase activity
positive regulation of triglyceride catabolic process
positive regulation of very-low-density lipoprotein particle remodeling
negative regulation of very-low-density lipoprotein particle clearance
lipid catabolic process
negative regulation of cholesterol transport
cholesterol efflux
phospholipid efflux
triglyceride-rich lipoprotein particle remodeling
chylomicron remodeling
very-low-density lipoprotein particle remodeling
chylomicron remnant clearance
high-density lipoprotein particle clearance
lipoprotein metabolic process
cholesterol homeostasis
negative regulation of catalytic activity
reverse cholesterol transport
positive regulation of fatty acid biosynthetic process
negative regulation of lipid metabolic process
negative regulation of receptor-mediated endocytosis
positive regulation of lipoprotein lipase activity
positive regulation of phospholipid catabolic process
triglyceride homeostasis
lipid binding
phospholipase activator activity
protein homodimerization activity
phospholipase binding
lipase inhibitor activity
lipoprotein lipase activator activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19008845.hg.1 | ENST00000252490.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008845.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008845.hg.1 | TC19002642.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008844.hg.1 | ENST00000252490.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008844.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008844.hg.1 | TC19002642.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008842.hg.1 | ENST00000252490.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008842.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008842.hg.1 | TC19002642.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011762.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201630.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201630.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201630.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201630.hg.1 | TC1900011763.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201630.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201630.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201630.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011763.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201614.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201614.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201614.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201614.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201614.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201614.hg.1 | TC1900011762.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201614.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201614.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201614.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201614.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201614.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201606.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201606.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201606.hg.1 | TC1900011762.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201606.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201606.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | TC1900011762.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | ENST00000252490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | ENST00000585786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | ENST00000590360.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | ENST00000591597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | ENST00000592257.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201613.hg.1 | NM_000483.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201629.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201629.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201629.hg.1 | NR_037932.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201629.hg.1 | TC1900011763.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201629.hg.1 | ENST00000585685.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201629.hg.1 | ENST00000589057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201629.hg.1 | NR_037932.1 | Refseq |

APOC2
Genomic Map