Assay Details
| Gene Symbol: LAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002294.2 | NP_002285.1 | 8 - 9 | 1278 | 84 | |
| GenBank mRNA | AK291090.1 | - | 8 - 9 | 1225 | 84 | |
| J04183.1 | - | 8 - 9 | 1190 | 84 | ||
| X77196.1 | - | 8 - 9 | 1235 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001325.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017774.hg.1 | ENST00000200639.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017774.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017774.hg.1 | TC0X001325.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00172535.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00172535.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00172535.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00172535.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00172535.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00172535.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00172535.hg.1 | TC0X00010671.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000200639.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000371335.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000434600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | ENST00000486593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_001122606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_002294.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010671.hg.1 | NM_013995.2 | Refseq |

LAMP2
Genomic Map