Assay Details
| Gene Symbol: POMC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000939.3 | NP_000930.1 | 1 - 2 | 358 | 70 | |
| GenBank mRNA | CB988233.1 | - | 1 - 2 | 432 | 70 | |
| CD108235.1 | - | 1 - 2 | 81 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_000939.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
generation of precursor metabolites and energy
signal transduction
neuropeptide signaling pathway
cell-cell signaling
regulation of blood pressure
peptide hormone processing
regulation of appetite
negative regulation of tumor necrosis factor production
cellular pigmentation
glucose homeostasis
positive regulation of transcription from RNA polymerase II promoter
regulation of glycogen metabolic process
regulation of corticosterone secretion
G-protein coupled receptor binding
receptor binding
hormone activity
neuropeptide hormone activity
type 3 melanocortin receptor binding
type 4 melanocortin receptor binding
type 1 melanocortin receptor binding
generation of precursor metabolites and energy
signal transduction
neuropeptide signaling pathway
cell-cell signaling
regulation of blood pressure
peptide hormone processing
regulation of appetite
negative regulation of tumor necrosis factor production
cellular pigmentation
glucose homeostasis
positive regulation of transcription from RNA polymerase II promoter
regulation of glycogen metabolic process
regulation of corticosterone secretion
G-protein coupled receptor binding
receptor binding
hormone activity
neuropeptide hormone activity
type 3 melanocortin receptor binding
type 4 melanocortin receptor binding
type 1 melanocortin receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000264708.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000380794.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000395826.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000395826.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000395826.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000395826.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000395826.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000395826.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000395826.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000395826.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000395826.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000395826.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000395826.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000405623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000449220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000449220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000449220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000449220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000449220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000449220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000449220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | ENST00000449220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_000939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011998.hg.1 | NM_001035256.2 | Refseq |

POMC
Genomic Map