| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027036.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027036.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027036.hg.1 |
TC02001680.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027038.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027038.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02027038.hg.1 |
TC02001680.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012071.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012071.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012071.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012071.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012071.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012071.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012071.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012071.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012071.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012071.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012071.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012071.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012071.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012071.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012071.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012071.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012071.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012071.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187989.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187989.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187989.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187989.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187989.hg.1 |
TC0200012071.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200187991.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187991.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187991.hg.1 |
ENST00000296099.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200187991.hg.1 |
NM_003353.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200187991.hg.1 |
TC0200012071.hg.1 |
Clariom D TC |