Assay Details
| Gene Symbol: CNKSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297647.1 | NP_001284576.1 | 1 - 2 | 109 | 72 | |
| NM_001297648.1 | NP_001284577.1 | 1 - 2 | 109 | 72 | ||
| NM_006314.2 | NP_006305.2 | 1 - 2 | 109 | 72 | ||
| GenBank mRNA | AB209335.1 | - | 1 - 2 | 104 | 72 | |
| AF100153.1 | - | 1 - 2 | 87 | 72 | ||
| AK001900.1 | - | 1 - 2 | 88 | 72 | ||
| AK222817.1 | - | 1 - 2 | 88 | 72 | ||
| AK222904.1 | - | 1 - 2 | 92 | 72 | ||
| AK292646.1 | - | 1 - 2 | 91 | 72 | ||
| AK296814.1 | - | 1 - 2 | 109 | 72 | ||
| AK302056.1 | - | 1 - 2 | 90 | 72 | ||
| BC011604.2 | - | 1 - 2 | 90 | 3139227 | 72 | |
| BC012797.2 | - | 1 - 2 | 90 | 3505293 | 72 | |
| BT006900.1 | - | 1 - 2 | 51 | 72 | ||
| DA445069.1 | - | 1 - 2 | 109 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000336.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001297648.1 transcripts.
Set Membership: |
|

CNKSR1
Genomic Map