| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21002927.hg.1 |
ENST00000355480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21002927.hg.1 |
ENST00000359759.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21002927.hg.1 |
ENST00000400337.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21002927.hg.1 |
ENST00000400347.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21002927.hg.1 |
ENST00000539645.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21002927.hg.1 |
NM_030582.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21002927.hg.1 |
NM_130445.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21002927.hg.1 |
TC21000245.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
ENST00000342220.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
ENST00000355480.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
ENST00000359759.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
ENST00000400337.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
NM_030582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
NM_130444.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
NM_130445.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
TC2100007420.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
ENST00000342220.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
ENST00000355480.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
ENST00000359759.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
ENST00000400337.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
NM_030582.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
NM_130444.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100150252.hg.1 |
NM_130445.3 |
Refseq |