| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018440.hg.1 |
ENST00000250018.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018440.hg.1 |
ENST00000341556.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018440.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11018440.hg.1 |
TC11003436.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000525406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000525406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000525406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000525406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000525406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000525406.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013150.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203833.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203833.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203833.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203833.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203833.hg.1 |
ENST00000250018.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203833.hg.1 |
ENST00000417164.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203833.hg.1 |
ENST00000528338.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100203833.hg.1 |
NM_004179.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100203833.hg.1 |
TC1100013150.hg.1 |
Clariom D TC |