Assay Details
| Gene Symbol: EIF1AY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278612.1 | NP_001265541.1 | 1 - 2 | 176 | 73 | |
| NM_004681.3 | NP_004672.2 | 1 - 2 | 176 | 73 | ||
| GenBank mRNA | AF000987.1 | - | 1 - 2 | 147 | 73 | |
| BC005248.1 | - | 1 - 2 | 58 | 3933718 | 73 | |
| BF694967.1 | - | 163 | 73 | |||
| BP331805.1 | - | 1 - 2 | 176 | 73 | ||
| DQ786267.1 | - | 1 - 2 | 99 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0Y000072.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF000987.1,BC005248.1,NM_001278612.1,NM_004681.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000485584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000485584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000485584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | ENST00000485584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000828.hg.1 | ENST00000361365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000828.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000828.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000828.hg.1 | TC0Y000072.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000827.hg.1 | ENST00000361365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000827.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000827.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000827.hg.1 | TC0Y000072.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000829.hg.1 | ENST00000361365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000829.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000829.hg.1 | TC0Y000072.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000825.hg.1 | ENST00000361365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000825.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000825.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000825.hg.1 | TC0Y000072.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000823.hg.1 | ENST00000361365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000823.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000823.hg.1 | TC0Y000072.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000826.hg.1 | ENST00000361365.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000826.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000826.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000826.hg.1 | TC0Y000072.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0Y00145800.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145800.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145800.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145800.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145800.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145800.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145800.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145800.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145800.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145800.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145800.hg.1 | TC0Y00006730.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000464196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000485584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000485584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000485584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | ENST00000485584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006730.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145799.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145799.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145799.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145799.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145799.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145799.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145799.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145799.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145799.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145799.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145799.hg.1 | TC0Y00006730.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0Y00145803.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145803.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145803.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145803.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145803.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145803.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145803.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145803.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145803.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145803.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145803.hg.1 | TC0Y00006730.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0Y00145797.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145797.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145797.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145797.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145797.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145797.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145797.hg.1 | TC0Y00006730.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0Y00145802.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145802.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145802.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145802.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145802.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145802.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145802.hg.1 | ENST00000382772.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145802.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145802.hg.1 | NM_001278612.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145802.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145802.hg.1 | TC0Y00006730.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0Y00145804.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145804.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145804.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145804.hg.1 | ENST00000361365.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145804.hg.1 | ENST00000465253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0Y00145804.hg.1 | NM_004681.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0Y00145804.hg.1 | TC0Y00006730.hg.1 | Clariom D TC |

EIF1AY
Genomic Map