Assay Details
| Gene Symbol: ITM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004867.4 | NP_004858.1 | 1 - 2 | 453 | 114 | |
| GenBank mRNA | AF038953.1 | - | 1 - 2 | 255 | 114 | |
| AK223327.1 | - | 1 - 2 | 232 | 114 | ||
| AY359051.1 | - | 1 - 2 | 194 | 114 | ||
| BC040437.1 | - | 1 - 2 | 208 | 5748519 | 114 | |
| BG506587.1 | - | 1 - 2 | 114 | |||
| CR456875.1 | - | 1 - 2 | 116 | 114 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001180.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015898.hg.1 | ENST00000373298.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015898.hg.1 | ENST00000469541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015898.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015898.hg.1 | TC0X001180.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000461357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000461357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000461357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169280.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169280.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169280.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169280.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169280.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169280.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169280.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169280.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169280.hg.1 | TC0X00010155.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000373298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000434584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000461357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000461357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000461357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000462038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000469541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | ENST00000482194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_001171581.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010155.hg.1 | NM_004867.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 171124 | - | - |

ITM2A
Genomic Map