| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050196.hg.1 |
ENST00000368736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050196.hg.1 |
ENST00000368737.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050196.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050196.hg.1 |
TC01003260.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050197.hg.1 |
ENST00000368736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050197.hg.1 |
ENST00000368737.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050197.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050197.hg.1 |
TC01003260.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015856.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015856.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015856.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015856.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015856.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015856.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015856.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015856.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015856.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015856.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015856.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015856.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015856.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015856.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015856.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015856.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015856.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015856.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219097.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219097.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219097.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219097.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219097.hg.1 |
TC0100015856.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100219095.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219095.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219095.hg.1 |
ENST00000368737.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219095.hg.1 |
NM_005621.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219095.hg.1 |
TC0100015856.hg.1 |
Clariom D TC |