Assay Details
| Gene Symbol: GLYAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005838.3 | NP_005829.3 | 1 - 2 | 142 | 62 | |
| NM_201648.2 | NP_964011.2 | 1 - 2 | 142 | 62 | ||
| GenBank mRNA | AB013093.1 | - | 1 - 2 | 91 | 62 | |
| AB665285.1 | - | 1 - 2 | 120 | 62 | ||
| AF023466.1 | - | 1 - 2 | 71 | 62 | ||
| BC009785.1 | - | 1 - 2 | 146 | 4045672 | 62 | |
| BG401676.1 | - | 1 - 2 | 182 | 62 | ||
| BG617408.1 | - | 1 - 2 | 124 | 62 | ||
| BP277146.1 | - | 1 - 2 | 143 | 62 | ||
| CB161978.1 | - | 1 - 2 | 66 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001813.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB665285.1,AF023466.1,BC009785.1,NM_005838.3,NM_201648.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glycine metabolic process
acyl-CoA metabolic process
xenobiotic metabolic process
response to toxic substance
monocarboxylic acid metabolic process
protein binding
transferase activity, transferring acyl groups
glycine N-acyltransferase activity
glycine N-benzoyltransferase activity
glycine metabolic process
acyl-CoA metabolic process
xenobiotic metabolic process
response to toxic substance
monocarboxylic acid metabolic process
protein binding
transferase activity, transferring acyl groups
glycine N-acyltransferase activity
glycine N-benzoyltransferase activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11020960.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020960.hg.1 | ENST00000344743.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020960.hg.1 | ENST00000529732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020960.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020960.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020960.hg.1 | TC11001813.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020961.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020961.hg.1 | ENST00000344743.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020961.hg.1 | ENST00000529732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020961.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020961.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020961.hg.1 | TC11001813.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000531084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000531084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000531084.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000586098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000586098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000586098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020963.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020963.hg.1 | ENST00000344743.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020963.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020963.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11020963.hg.1 | TC11001813.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000531084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000531084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000531084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000586098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000586098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000586098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010948.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182647.hg.1 | TC1100010948.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | ENST00000531084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | ENST00000529732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | ENST00000531084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | ENST00000611865.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182649.hg.1 | TC1100010948.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100182653.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182653.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182653.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182653.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182653.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182653.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182653.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182653.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182653.hg.1 | TC1100010948.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100182652.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182652.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182652.hg.1 | ENST00000531084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182652.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182652.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182652.hg.1 | ENST00000278400.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182652.hg.1 | ENST00000344743.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182652.hg.1 | ENST00000531084.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100182652.hg.1 | NM_005838.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182652.hg.1 | NM_201648.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100182652.hg.1 | TC1100010948.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 202611 | - | - |

GLYAT
Genomic Map