| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15008497.hg.1 |
ENST00000268164.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15008497.hg.1 |
ENST00000539113.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15008497.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15008497.hg.1 |
TC15000883.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1500160707.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500160707.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500160707.hg.1 |
ENST00000555434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500160707.hg.1 |
ENST00000556382.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500160707.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500160707.hg.1 |
TC1500008370.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500160707.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500160707.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500160707.hg.1 |
ENST00000555434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500160707.hg.1 |
ENST00000556382.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500160707.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000268164.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000539113.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000555434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
ENST00000556382.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008370.hg.1 |
NM_006011.3 |
Refseq |