Assay Details
| Gene Symbol: S100A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006271.1 | NP_006262.1 | 1 - 2 | 104 | 74 | |
| GenBank mRNA | AK312152.1 | - | 1 - 2 | 61 | 74 | |
| BC014392.1 | - | 1 - 2 | 66 | 3543900 | 74 | |
| X58079.1 | - | 1 - 2 | 104 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001258.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312152.1,BC014392.1,NM_006271.1,X58079.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of heart contraction
substantia nigra development
intracellular signal transduction
positive regulation of voltage-gated calcium channel activity
calcium ion binding
protein binding
identical protein binding
protein homodimerization activity
S100 protein binding
calcium-dependent protein binding
ATPase binding
negative regulation of transcription from RNA polymerase II promoter
regulation of heart contraction
substantia nigra development
intracellular signal transduction
positive regulation of voltage-gated calcium channel activity
calcium ion binding
protein binding
identical protein binding
protein homodimerization activity
S100 protein binding
calcium-dependent protein binding
ATPase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01019180.hg.1 | ENST00000292169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019180.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019180.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019180.hg.1 | ENST00000469893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019180.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019180.hg.1 | TC01001258.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019175.hg.1 | ENST00000292169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019175.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019175.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019175.hg.1 | ENST00000469893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019175.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019175.hg.1 | TC01001258.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000436839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000436839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000436839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019176.hg.1 | ENST00000292169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019176.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019176.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019176.hg.1 | ENST00000469893.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019176.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01019176.hg.1 | TC01001258.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | ENST00000436839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | ENST00000436839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172746.hg.1 | TC0100010043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000436839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000436839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000436839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010043.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | ENST00000436839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | ENST00000436839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172741.hg.1 | TC0100010043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100172740.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172740.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172740.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172740.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172740.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172740.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172740.hg.1 | ENST00000368696.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172740.hg.1 | ENST00000368698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172740.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172740.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172740.hg.1 | TC0100010043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100172737.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172737.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172737.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172737.hg.1 | ENST00000292169.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172737.hg.1 | ENST00000469893.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172737.hg.1 | NM_006271.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172737.hg.1 | TC0100010043.hg.1 | Clariom D TC |

S100A1
Genomic Map