Assay Details
| Gene Symbol: HNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006895.2 | NP_008826.1 | 2 - 3 | 441 | 107 | |
| XM_017003949.1 | XP_016859438.1 | 2 - 3 | 274 | 107 | ||
| GenBank mRNA | AF523358.1 | - | 2 - 3 | 368 | 107 | |
| AF523359.1 | - | 2 - 3 | 368 | 107 | ||
| AF523360.1 | - | 2 - 3 | 368 | 107 | ||
| AK301462.1 | - | 2 - 3 | 199 | 107 | ||
| BC020677.1 | - | 2 - 3 | 200 | 4767331 | 107 | |
| D16224.1 | - | 2 - 3 | 197 | 107 | ||
| U08092.1 | - | 2 - 3 | 226 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000891.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
histamine catabolic process
response to tumor cell
histidine catabolic process
hyperosmotic response
brain development
respiratory gaseous exchange
response to amine
methylation
response to immobilization stress
response to cocaine
response to glucocorticoid
response to interleukin-1
positive regulation of protein targeting to mitochondrion
histamine N-methyltransferase activity
histamine catabolic process
response to tumor cell
histidine catabolic process
hyperosmotic response
brain development
respiratory gaseous exchange
response to amine
methylation
response to immobilization stress
response to cocaine
response to glucocorticoid
response to interleukin-1
positive regulation of protein targeting to mitochondrion
histamine N-methyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000329366.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000329366.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000329366.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000329366.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000329366.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000467390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000467390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000467390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000467390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000467390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000475675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000475675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000475675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000475675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000475675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000480534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000480534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000480534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_001024074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_001024074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_001024074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_001024074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_001024074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_001024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_001024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_001024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_001024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_001024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_001024075.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013224.hg.1 | ENST00000280097.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013224.hg.1 | ENST00000410115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013224.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013224.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013224.hg.1 | TC02000891.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000329366.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000329366.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000329366.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000329366.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000329366.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000467390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000467390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000467390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000467390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000467390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000475675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000475675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000475675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000475675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000475675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000480534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000480534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000480534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_001024074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_001024074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_001024074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_001024074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_001024074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_001024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_001024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_001024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_001024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_001024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_001024075.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009470.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200166339.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200166339.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200166339.hg.1 | ENST00000467390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200166339.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200166339.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200166339.hg.1 | ENST00000280097.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200166339.hg.1 | ENST00000410115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200166339.hg.1 | ENST00000467390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200166339.hg.1 | ENST00000485653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200166339.hg.1 | NM_006895.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200166339.hg.1 | TC0200009470.hg.1 | Clariom D TC |

HNMT
Genomic Map