Assay Details
| Gene Symbol: MYRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013279.3 | NP_037411.1 | 1 - 2 | 57 | 56 | |
| GenBank mRNA | AF086762.1 | - | 1 - 2 | 57 | 56 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000539.hg
Additional Information:
This assay is in the 5'UTR region of the AF086762.1,NM_013279.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
proteolysis
oligodendrocyte development
central nervous system myelination
positive regulation of myelination
central nervous system myelin maintenance
positive regulation of transcription, DNA-templated
oligodendrocyte differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
peptidase activity
transcription, DNA-templated
proteolysis
oligodendrocyte development
central nervous system myelination
positive regulation of myelination
central nervous system myelin maintenance
positive regulation of transcription, DNA-templated
oligodendrocyte differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
peptidase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000265460.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000278836.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000389602.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000536352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000537766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000539361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | ENST00000546247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_001127392.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007819.hg.1 | NM_013279.3 | Refseq |

MYRF
Genomic Map