Assay Details
| Gene Symbol: N4BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153029.3 | NP_694574.3 | 2 - 3 | 2128 | 90 | |
| XM_011523482.1 | XP_011521784.1 | 2 - 3 | 2133 | 90 | ||
| GenBank mRNA | AB014515.1 | - | 2 - 3 | 2128 | 90 | |
| AK001464.1 | - | 1 - 2 | 538 | 90 | ||
| AK026937.1 | - | 1 - 2 | 1219 | 90 | ||
| BC108288.1 | - | 1 - 2 | 1689 | 90 | ||
| BC152472.1 | - | 2 - 3 | 2128 | 8862609 | 90 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001098.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16015438.hg.1 | ENST00000262384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015438.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015438.hg.1 | TC16001098.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015439.hg.1 | ENST00000262384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015439.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015439.hg.1 | TC16001098.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015440.hg.1 | ENST00000262384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015440.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015440.hg.1 | TC16001098.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000564710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000564710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000565423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000565423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000565423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000565423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000565638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000565638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000565638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000569027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000569027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000569027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | ENST00000569027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015441.hg.1 | ENST00000262384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015441.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015441.hg.1 | TC16001098.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000564710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000564710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000565423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000565423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000565423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000565423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000565638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000565638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000565638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000569027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000569027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000569027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | ENST00000569027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010227.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178775.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178775.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178775.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178775.hg.1 | TC1600010227.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600178775.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178775.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178775.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178776.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178776.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178776.hg.1 | ENST00000564710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178776.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178776.hg.1 | TC1600010227.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600178776.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178776.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178776.hg.1 | ENST00000564710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178776.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178772.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178772.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178772.hg.1 | ENST00000565638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178772.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178772.hg.1 | TC1600010227.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600178772.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178772.hg.1 | ENST00000564124.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178772.hg.1 | ENST00000565638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178772.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178778.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178778.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178778.hg.1 | TC1600010227.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600178778.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178778.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178780.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178780.hg.1 | NM_153029.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178780.hg.1 | TC1600010227.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600178780.hg.1 | ENST00000262384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178780.hg.1 | NM_153029.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 101887 | - | - |

N4BP1
Genomic Map